Order Entry
United States
ContactUsLinkComponent
EZ-Accu Shot™ Burkholderia multivorans derived from ATCC® BAA-247™, Microbiologics
EZ-Accu Shot™ Burkholderia multivorans derived from ATCC® BAA-247™, Microbiologics
Catalog # 77394-844
CAS Number:  
EZ-Accu Shot™ Burkholderia multivorans derived from ATCC® BAA-247™, Microbiologics
Catalog # 77394-844
Supplier Number:  01270A
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Burkholderia multivorans derived from ATCC® BAA-247™
  • Size:
    1 kit
  • Cat. No.:
    77394-844

Specifications

About this item

Microorganisms, Microbiologics to EZ-Accu Shot™

  • Quantitative microorganism preparation that delivers less than 100 CFU per 0.1 ml of hydrated suspension
  • No dilutions required
  • Growth Promotion Test
  • Taxomy: Bacteria

Kit contains: 5 pellets of a single enumerated strain and 5 vials of hydrating fluid (1.2 ml per vial)