Order Entry
United States
ContactUsLinkComponent
Anti-Influenza HA Neutralizing (H5-specific) Monoclonal Antibody [clone: RV28]
Anti-Influenza HA Neutralizing (H5-specific) Monoclonal Antibody [clone: RV28]
Supplier:  ReVacc Scientific
Certificates
undefined
Anti-Influenza HA Neutralizing (H5-specific) Monoclonal Antibody [clone: RV28]
Supplier:  ReVacc Scientific

Specifications

  • Catalog No:
  • 77394-682
  • 77394-680
  • Antigen Name:
  • Influenza A (H5)
  • Influenza A (H5)
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • RV28
  • RV28
  • Reactivity:
  • Human
  • Human
  • Isotype:
  • IgG1
  • IgG1
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Epitope:
  • Influenza A HA
  • Influenza A HA
  • Form:
  • Liquid in PBS, pH7.2, without preservatives
  • Liquid in PBS, pH7.2, without preservatives
  • Purification:
  • Cell culture supernatant followed by filtration and Protein A purification
  • Cell culture supernatant followed by filtration and Protein A purification
  • Concentration:
  • 1 mg/mL by Refractometry
  • 1 mg/mL by Refractometry
  • Storage Buffer:
  • PBS, pH 7,2.
  • PBS, pH 7,2.
  • Storage Temperature:
  • Store at 4 °C, -20°C
  • Store at 4 °C, -20°C
  • Shipping Temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together