Human IgE (Non-Immune, Azide)
Catalog # 76529-708
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Conjugation:Unconjugated
- Species:Human
- Size:400 µg
- Protein Synonyms:Immunoglobulin E
- Protein/Peptide Name:IgE (non-immune, azide)
- Formulation:Liquid. In 0.01 M phosphate buffer, pH 7.4, containing 0.15 M sodium chloride and 0.09% sodium azide
- Shipping Temperature:20
- Cat. No.:76529-708
Specifications
About this item
Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma.