Recombinant Coronavirus (SARS-CoV NP) Antigen (aa 340-390) (from E. coli)
76418-084
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:100 µg
- Storage Conditions:–20 °C
- Protein Synonyms:Coronavirus nucleoprotein
- Protein/Peptide Name:Coronavirus (SARS-CoV NP) antigen (aa 340-390)
- Molecular Weight:32 kDa
- Formulation:Liquid. In 50 mM Tris-HCl, 60 mM NaCl with 50% glycerol.
- Shipping Temperature:20
- Cat. No.:76418-084
Produced in E. coli. C-terminal coronavirus (SARS-CoV) nucleoprotein (aa 340-390).