Recombinant Coronavirus (SARS-CoV Matrix) Antigen (aa 182-216) (from E. coli)
Catalog # 76418-076
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Size:100 µg
- Storage Conditions:–20 °C
- Protein Synonyms:Coronavirus matrix protein
- Protein/Peptide Name:Coronavirus (SARS-CoV Matrix) antigen (aa 182-216)
- Molecular Weight:30 kDa
- Formulation:Liquid. In 50 mM Tris-HCl, 60 mM NaCl with 50% glycerol.
- Shipping Temperature:20
- Cat. No.:76418-076
Specifications
About this item
Produced in E. coli. Coronavirus (SARS-CoV) matrix protein (aa 182-216).