Recombinant Coronavirus (COVID-19 S2) Antigen (aa 685-1211) (from Mammalian cells)
Catalog # 76418-070
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:Mammalian cells
- Species:
- Size:100 µg
- Environmentally Preferable:
- Storage Conditions:–80 °C
- Protein Synonyms:Coronavirus spike protein
- Protein/Peptide Name:Coronavirus (COVID-19 S2) antigen (aa 685-1211)
- Formulation:Liquid. In DPBS, pH 7.5.
- Shipping Temperature:Dry Ice
- Cat. No.:76418-070
Specifications
About this item
Produced in a mammalian cells expression system. Coronavirus (COVID-19) spike protein (aa 685-1211) is fused to a Fc-tag.