Human Recombinant BMPR-1A (from CHO cells)
Catalog # 76418-038
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:CHO cells
- Species:Human
- Size:20 µg
- Storage Conditions:–80 °C
- Endotoxin Content:≤1 EU/µg
- Reconstitution Instructions:Reconstitute in sterile water at 0.1 mg/ml
- Protein Synonyms:Activin receptor-like kinase 3|SKR5|BMP type-1A receptor
- Protein/Peptide Name:BMPR-1A
- Molecular Weight:Dimer, 40.4/80.8 kDa (362/724 amino acids)
- Formulation:Lyophilized from a sterile (0.2 µm) filtered aqueous solution containing PBS
- Shipping Temperature:Ambient temperature
- Cat. No.:76418-038
Specifications
About this item
Produced in genetically modified CHO cells. Contains a C-terminus linker (IEGRMD) and an Fc tag (Human IgG1).