Orders LinkContactUsLinkComponent
Boot Covers
Boot Covers
Blue, Universal
Catalog # 76478-458
Supplier:   Dukal
Supplier #:  355-15
Boot Covers
Catalog # 76478-458
Supplier:  Dukal
Boot Covers
Boot Covers
Blue, Universal
Price
$531.66
Log in for availability
Case of 20
Product Details & Documents
Boot covers protect the shoe and lower portion of the leg.
  • Ultrasonic sealed seams
  • Includes 2 black anti-slip strips on the bottom of each foot
  • Tie at ankle for a comfortable and adjustable fit
Constructed of SMS with a PE film coating for moderate coverage and fluid resistance.
Packaging: Boot covers are packaged 20 per bag, 15 bags per case.
Specifications
  • Clothing Size:
    Universal
  • Color:
    Blue