FitMe™ Lab Coats
Supplier: Dukal
Certificates
About this item
FitMe™ lab coats feature a unique adjustable waistband design that provides a true custom fit.
- Knit cuffs and collar
- Snap front closure
- Premium SMS material
- 2 hip pockets, 1 chest pocket
- Washable and autoclavable up to 5 times
Available in a wide variety of colors, FitMe™ lab coats are manufactured from premium SMS material, and are supplied non sterile.
Packaging: Lab coats are packaged 10 per bag, 5 bags per case.
Specifications
- Catalog No:
- 76582-256
- 76582-198
- 76582-180
- 76582-166
- 76582-156
- 76582-278
- 76582-114
- 76582-232
- 76582-182
- 76582-220
- 76582-200
- 76582-262
- 76582-224
- 76582-190
- 76582-186
- 76582-282
- 76582-192
- 76582-248
- 76582-160
- 76582-228
- 76582-210
- 76582-272
- 76582-134
- 76582-268
- 76582-208
- 76582-162
- 76582-168
- 76582-112
- 76582-196
- 76582-188
- 76582-242
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Closure type:
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- With Press-Stud Fastening
- Fabric:
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS
- Premium SMS