Spunbond Bouffant Cap
Catalog # 77322-744
Supplier: O&M HALYARD, INC
Specifications
- Clothing Size:L
- Color:Blue
- Description:Spunbond bouffant caps
- Cat. No.:77322-744
Specifications
About this item
Spunbond bouffant caps are made with lightweight spunbond fabric and are designed to cover hair in 'no fluid contamination' or 'light fluid contamination' environments.
- Single-layer spunbond fabric
- Latex-free elastic
- No potential fluid contact
Spunbond bouffant caps are available in various sizes and colors to meet different needs at healthcare facilities. These caps are suitable for use in non-fluid contamination environments, such as patient rooms, waiting areas, and exam rooms. They are roomy and lightweight to wear, keeping hair securely tucked away.