ContactUsLinkComponent
Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
Catalog # 60004-040
Supplier:  Kemtech America
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
undefined
Synthware High Vacuum Valve, Teflon® Protected, 3-Way Metering
Catalog # 60004-040
Supplier:  Kemtech America
Supplier Number:  S640008
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
Specifications
  • Description:
    High vacuum valve, Teflon® protected, 3-way metering
  • Cat. No.:
    60004-040
  • Valve size:
    0 - 8 mm
  • Supplier no.:
    S640008
Specifications
About this item
Viton® O-rings on PTFE shaft are protected with precision Teflon® wipers. The plug has a machined-in integral PTFE tip O-ring to make the primary seal.
  • Teflon® shaft
  • PTFE covered Viton® O-rings
Valves are designed to provide an excellent vacuum tight seal without exposing any material other than Teflon® to chemicals.