Order Entry
United States
ContactUsLinkComponent
Klinger Laser Ray Box, Electronic
Klinger Laser Ray Box, Electronic
  470346-482
 :  Klinger
undefined
Klinger Laser Ray Box, Electronic
  470346-482
 :  Klinger
 :  LRB-ELEC-KL

 

  • Description:
    Laser ray box, electronic
  • Cat. No.:
    470346-482

 

 

This laser ray box has several modes, with 1, 3, or 5 beams.

  • Multiple ray configurations options: five rays (1, 2, 3, 4, 5) - three rays (1, 3, 5) - three rays (2, 3, 4) - single ray (3)
  • Powered by 2×AA batteries or included 3 V/300 mA power supply
  • Laser type: diode wavelength, 635 nm
  • Class II Laser, <1 mW

The electronic laser ray box consists of five independent red laser modules producing five parallel lines that are collimated by cylindrical lenses to be highly visible. This electronic version adds electronic shade control for each of the beams. It is taped with magnetized foil, allowing intuitive attachment to a magnetic board. Use the ON/OFF switch to select between one ray, three central rays, three alternating rays, or all five rays depending on specific needs.