Orders LinkContactUsLinkComponent
Eisco Laser Ray Box
Eisco Laser Ray Box
Laser ray box
Catalog # 470346-456
Supplier:   Wards
Supplier #:  LRBX
Eisco Laser Ray Box
Catalog # 470346-456
Supplier:  Wards
Eisco Laser Ray Box
Eisco Laser Ray Box
Laser ray box
Price
$177.00
Log in for availability
Each
Product Details & Documents
This laser ray box is adjustable, magnetic, and bright enough to use in ambient light.
  • Operates on 5 V USB power supply or rechargeable battery
  • Produces either 3 or 5 parallel laser beams
  • Bright enough to use in a classroom setting with ambient light
  • Magnetic strip included for whiteboard use
When the traditional white light ray boxes do not provide strong enough rays to be seen across a classroom of ambient light, then we recommend this 635 nm wavelength, 3/5 ray, monochromatic (laser) ray box. The battery powered ray box comes with a USB cable so it can easily be charged with a computer, a charging station, or a standard USB wall adapter. An included magnetic strip may be applied to the back of the ray box so that it can be used with a metalic white board for demonstrations. Easily toggle between three or five parallel laser rays.
Specifications
  • Description:
    Laser ray box