Automation Tips for Agilent® Bravo and VPrep® Workstations
Fluotics manufactures a complete selection of automation tips compatible with the Agilent™ Bravo and VPrep. The automation tips are directly interchangeable with their Agilent robotic tip counterpart with no change to the labware definitions required.
- Certified RNase-/DNase-free and nonpyrogenic
- Produced from virgin, high grade polypropylene
- Available in non-sterile, sterile and barrier filter formats
Compatible with Agilent™ VPrep® or Bravo® 96 or 384 ST series (Short tip, type II or III) multi-channels heads. Ultra polishing techniques ensure low liquid retention without the use of releasing agents or other additives.
: Packaged in 96 or 384 tips per rack, 50 racks per case.
Frequently Bought Together