Plastic Dropper
Dropper, plastic, pack of 12, 1 ml, 3"
Catalog # 470352-216
Supplier: United Scientific Supplies
Supplier #: PLPIP3
Jump to:
Product Details & Documents
Economical plastic dropper.
- PP tube
- Rubber bulb
Straight plastic pipettes with a capcity of 1 ml. The tips are PP and the bulb is made out of rubber.
Specifications
- Capacity:1 ml
- Description:Dropper, plastic, pack of 12
- Overall length:3"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



