Sprint™ 8 Plus Clinical Centrifuge, 230 V
Catalog # 470334-152
Supplier: Ward's
Irregular Voltage
Specifications
- Max. Capacity:8×5/7 ml
- Max. Speed:200 - 6800 rpm
- Description:Sprint™ 8 plus clinical centrifuge, EU-plug
- Electrical:230 V, 50/60 Hz, 100 W
- Max. RCF:10 - 4445 ×g
- Dimensions:280×350×95 mm (11×13.7×9.5")
- Timer:Up to 59 hours, 99 minutes
- Weight:8 kg (17.6 lbs.)
- Cat. No.:470334-152
Specifications
About this item
The Sprint™ 8 PLUS allows for the separation of serum or plasma in as little as 2 to 3 minutes.
- 230 V model, not for use in USA
- Unique EZ-Scroll™ touch pad
- Ideal for rapid production of serum and plasma
- Program storage up to 99
- Adjustable speed and timer
- 8 Position fixed angle rotor rotor
- Digital speed control in rpm or rcf with adjustable acceleration and deceleration rates (up to 6800 rpm/4445 ×g)
Additional features includes, A large back-lit, liquid crystal display, High capacity rotor, 8 tubes up to 15 ml (conical or round bottom), Timer up to 99 hr 99 mins, Storage of commonly used run-times, Control of speed in in both rpm and rcf, Adjustable acceleration and deceleration levels, Safety imbalance detection system, Momentary operation and Electrical lid lock.
Speed increment - 10 rpm or 10 rcf
Caution: 230 V model, not for use in USA. Corresponding model with voltage suitable for use in US (470334-150) is also available, please see separate product page.