ContactUsLinkComponent
Staticide® Bench Mount Grounding Station for Two Wrist Straps
Staticide® Bench Mount Grounding Station for Two Wrist Straps
Catalog # 77200-572
Supplier:  ACL Staticide
undefined
Staticide® Bench Mount Grounding Station for Two Wrist Straps
Catalog # 77200-572
Supplier:  ACL Staticide
Supplier Number:  8092
Specifications
  • Description:
    Bench mount grounding station for two wrist straps
  • Cat. No.:
    77200-572
Specifications
About this item
Connect the 8092 bench mount ground cord to an earth ground.
  • 10' coated high-strength tensile wire
  • 1-meg ohm resistor
  • Attach under workbench table top
  • Ring-style termination for connecting to ground point
  • Equipped with two banana jack plug-ins
Cable is equippped with 1-megohm resistor. Parking system attaches under an ESD workstation and offers two banana plug-in points for wrist straps.