Staticide® Pres-N-Snap Installation Hand Tool
Catalog # 77200-608
Supplier: ACL Staticide
Specifications
- Description:Pres-N-Snap Tool for grounding ESD mats
- Material:Cast aluminum
- Cat. No.:77200-608
Specifications
About this item
Heavy-duty clamping tool for attaching grounding point snaps on ESD-safe dissipative mats.
- Heavy-duty cast aluminum construction
- Made in USA
Includes attaching dies for 1 cm snaps, and ten pairs of female sockets and eyelets.