Order Entry
United States
ContactUsLinkComponent
Diluent A (with rAlbumin)
Diluent A (with rAlbumin)
Catalog # 77322-302
CAS Number:  
Diluent A (with rAlbumin)
Catalog # 77322-302
Supplier Number:  B8532S
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Concentration:
    1X
  • Description:
    Diluent A (with rAlbumin)
  • Size:
    5 ml
  • Cat. No.:
    77322-302

Specifications

About this item

Diluent A (with rAlbumin) is recommended for making dilutions of New England Biolabs, Inc. restriction enzymes.

  • Contains 4×1.25 ml vials
  • Restriction enzymes should be stored at their selling concentrations
  • Contains rAlbumin

Diluent A (with rAlbumin) is recommended for making dilutions of some NEB restriction enzymes. When necessary, we recommend diluting enzymes just prior to use and suggest that the final concentration of diluted enzymes be at least 1000 units/ml. Restriction enzymes should be stored at their selling concentrations.