Orders LinkContactUsLinkComponent
Digital Water Baths
Digital Water Baths
Digital Water Baths
Certificates
Digital Water Baths
4 Options Available Below
Product Details & Documents
Available in mutiple sizes, these elegant water baths have stainless steel chambers and LED displays.
  • Easy-touch glass panel interface
  • Adjustable temperature, ambient 5 to 99 °C
  • Dry-start protection
  • Customizable over-temperature cut-off and alarm
  • 2 and 5 L models have a 1 to 999 minute timer; 13 and 24 L models have a 0 to 99 hour and 59 minute timer
  • AC 100 - 120 V, 50/60 Hz
Temperature settings can be adjusted with a resolution of 0.1 °C, with great stability and uniformity. Users can switch between Fahrenheit and Celsius as needed. The 13 and 24 L models feature a drainage port for convenience.
Specifications for Product Family
Specifications
  • Catalog No:
  • 470354-082
  • 470354-084
  • 470354-080
  • 470354-086
  • Model:
  • WB101
  • WB201
  • WB301
  • WB401
  • Temperature Stability:
  • ±0.5 °C
  • ±0.5 °C
  • ±0.3 °C
  • ±0.3 °C
  • Temperature Uniformity:
  • ±0.5 °C
  • ±0.5 °C
  • ±0.5 °C
  • ±0.3 °C
  • Timer:
  • 1 - 999 minutes
  • 1 - 999 minutes
  • 0 - 99 h 59 min
  • 0 - 99 h 59 min
Specifications
Product Family Options
Product Information
  • Interior W×D×H
    Availability
    Price
  • 470354-082
    Single chamber, 2 L
    Single chamber, 2 L
    138×150×150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $299.00
    $299.00
    Each
       $299.00
    Minimum order quantity is 1
    Supplier Number: WB101V11-4EUSA
    Specifications:
    Irregular Voltage
    More
    Specifications
    Cat. No.
    470354-082
    Model
    WB101
    Temperature Stability
    ±0.5 °C
    Temperature Uniformity
    ±0.5 °C
    Timer
    1 - 999 minutes
    Irregular Voltage
  • 470354-084
    Dual chambers, 5 L
    Dual chambers, 5 L
    150×300×150 mm
    Log in for availability
    Log in for availability
    Each
    Each - $479.00
    $479.00
    Each
       $479.00
    Minimum order quantity is 1
    Supplier Number: WB201V11-4EUSA
    Specifications:
    Irregular Voltage
    More
    Specifications
    Cat. No.
    470354-084
    Model
    WB201
    Temperature Stability
    ±0.5 °C
    Temperature Uniformity
    ±0.5 °C
    Timer
    1 - 999 minutes
    Irregular Voltage
  • 470354-080
    Single chamber, 13 L
    Single chamber, 13 L
    300×240×200 mm
    Log in for availability
    Log in for availability
    Each
    Each - $879.00
    $879.00
    Each
       $879.00
    Minimum order quantity is 1
    Supplier Number: WB301V11-4EUSA
    Specifications:
    Irregular Voltage
    More
    Specifications
    Cat. No.
    470354-080
    Model
    WB301
    Temperature Stability
    ±0.3 °C
    Temperature Uniformity
    ±0.5 °C
    Timer
    0 - 99 h 59 min
    Irregular Voltage
  • 470354-086
    Dual chambers, 24 L
    Dual chambers, 24 L
    500×300×200 mm
    Log in for availability
    Log in for availability
    Each
    Each - $1,299.00
    $1,299.00
    Each
       $1,299.00
    Minimum order quantity is 1
    Supplier Number: WB401V11-4EUSA
    Specifications:
    Irregular Voltage
    More
    Specifications
    Cat. No.
    470354-086
    Model
    WB401
    Temperature Stability
    ±0.3 °C
    Temperature Uniformity
    ±0.3 °C
    Timer
    0 - 99 h 59 min
    Irregular Voltage