Human Recombinant WRN Protein (His Tag) (from HEK293 Cells)
Catalog # 77295-220
Supplier: Sino Biological
CAS Number:
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Size:20 µg
- Environmentally Preferable:
- Endotoxin Content:<1.0 EU per μg protein as determined by the LAL method
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:RECQL3|RECQ3|RECQL2
- Protein/Peptide Name:WRN
- Purity:≥90% as determined by SDS-PAGE.
- Cat. No.:77295-220
Specifications
About this item
A DNA sequence encoding the human WRN (Q14191) (Met1-Ile333) was expressed, with a polyhistidine tag at the N-terminus.