Order Entry
United States
ContactUsLinkComponent
Freezing Point Osmometer
Freezing Point Osmometer
Catalog # 76644-946
undefined
Freezing Point Osmometer
Catalog # 76644-946
Supplier Number:  OSM100100

Specifications

  • Range:
    0 - 3000 mOsm/kg
  • Description:
    Freezing point osmometer OSM100100, single, 23×21×36 cm
  • Sample Volume:
    60 μl
  • Test Time:
    <2 min 30 sec/run
  • Cat. No.:
    76644-946

Specifications

About this item

The OSM100100 Osmometer measures the total osmolality of any aqueous solution by Freezing Point Depression (FPD).

  • Built-in touchscreen for intuitive operation
  • Small sample quantity required for testing
  • No proprietary consumables required
  • Refrigerant-free dual cooling system
  • Simple calibration procedure using 2 or more points
  • Built-in printer for test results

Designed for clinical and laboratory analysis of aqueous solutions, the osmometer is suitable to test urine, serum, or other bodily fluids. No proprietary consumables are required with a 0.5 ml sample tube used as the sample holder. The instrument utilizes a highly precise sensor for accurate measurement and high repeatability. Molar concentration, freezing point, and molar concentration ratio are all measured and recorded.

The multipoint calibration feature guarantees accurately calibrated results, allowing from 2 up to 30 total calibration points. The touchscreen enables intuitive user operation. A built-in printer provides convenient output of test results, with a variety of formatting options available. Data storage, access management, and password login for administrative functions ensure that authorized users can view and manage test result data securely.

Accuracy: ±2 mOsm/kg (≤400 mOsm/kg), ±1.0% (>400 mOsm/kg)

Accessories information: The instrument uses a 0.5 ml sample tube. Four E’s sample tube or Axygen PCR Tube PCR-05-C are compatible.
Delivery: Supplied with an operating manual, inspection sheet, 1 pack of Four E's 0.5 ml sample tubes, rubber suction bulb, touch pen and a power cord.