Human Recombinant FGF18/FGF-18 Protein (His Tag) (from HEK293 Cells)
77295-104
:
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:1 mg
- Endotoxin Content:<1.0 EU per μg of the protein as determined by the LAL method
- Biological Activity:1. Measured by its binding ability in a functional ELISA.
2. Immobilized human FGF18 (Cat: 13206-H08H) at 10 μg/ml (100 μl/well) can bind Rat FGFR4 (Cat:80093-R02H), The EC50 of Rat FGFR4 (Cat:80093-R02H) is 1.17 μg/ml. - Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:ZFGF5|FGF-18
- Protein/Peptide Name:FGF18
- Purity:>92% as determined by SDS-PAGE.
- Cat. No.:77295-104
A DNA sequence encoding the human FGF18 (O76093) (Met 1-Ala 207) was fused with a polyhistidine tag at the C-terminus.