Human Recombinant IL20/Interleukin-20 Protein (from E. coli)
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the mature form of human IL20 (Q9NYY1) (Leu25-Glu176) was expressed, with a N-terminal Met.
Specifications
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Human
- Biological Activity:Measured in a cell proliferation assay using BaF3 mouse pro-B cells transfected with human IL20 Rα and human IL20 Rβ. The ED50 for this effect is typically 0.25 - 2.5 ng/ml.
- Endotoxin-free:No
- Carrier-Free:Yes
- Animal-Free:No
- Protein Synonyms:IL10D|interleukin-20|IL-20|ZCYTO10
- Protein/Peptide Name:IL20
- Purity:>95% as determined by SDS-PAGE.
Specifications
Frequently Bought Together