Human Recombinant CD86 Protein (His and AVI Tag) (from HEK293 Cells), Biotinylated
Supplier: Sino Biological
Certificates
About this item
A DNA sequence encoding the human CD86 (NP_008820.4) (Met1-His239) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the biotin-protein ligase (BirA enzyme) which is co-expressed.
Specifications
- Source:
- Species:
- Environmentally Preferable:
Specifications
Frequently Bought Together