About this item
Probe-based one step RT-qPCR 2X master mix contains Reverse Transcriptase, Taq DNA polymerase, low ROX reference dye, dNTPs and optimized buffer with stabilizers. This master mix is ideal for high-throughput real-time PCR screening and validation which requires minimal handling and offer robust and consistent results.
- Enhanced efficiency, specificity, and sensitivity
- Compatible with all real-time PCR instruments
- Superior gene expression results under various cycling conditions
Applications:
Gene expression analysis
Genotyping
Mutation detection, pathogen detection, viral detection and absolute quantification
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Application:
- Environmentally Preferable: