Ziath DataPaq™ Mirage
Supplier: Avantor
Certificates
About this item
Economical camera-based reader with small footprint and low height, fitted with USB output and cryoprotection for use with cold racks.
- Reads all 2D Datamatrix coded tubes
- Low profile for easy integration with liquid handling and other automation platforms
- Export data to Excel, Text, XML, JSON and image files
- Easy to set up and easy to use
- Offers easy, out-of-the-box set up and is delivered pre-calibrated and ready to scan
- With Cryoprotection: Prevents the scanner window from misting when scanning a cold rack from a freeze
- Our passive technology does not use heat or blown air
Software includes remote control capability for integration applications.
Operating system on host PC: Windows® 7, 8, 10.
Compatible with 24, 48, 96, 240, 384 SBS racks.
Certifications: CE marked.
Ordering information: Two-year warranty.
Packaging: One reader system per case.