Order Entry
United States
ContactUsLinkComponent
Zebrafish Conditioned Place Preference Chamber
Zebrafish Conditioned Place Preference Chamber
Catalog # 76670-108
Supplier:  MAZE ENGINEERS
undefined
Zebrafish Conditioned Place Preference Chamber
Catalog # 76670-108
Supplier:  MAZE ENGINEERS
Supplier Number:  4804
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Width:
    17 cm
  • Description:
    Conditioned place preference chamber, zebrafish
  • Height:
    23 cm
  • Length:
    52 cm
  • Material:
    Acrylic
  • Environmentally Preferable:
  • Cat. No.:
    76670-108

Specifications

About this item

The Zebrafish conditioned place preference (CPP) chamber is a widely used paradigm to evaluate the rewarding effects of food substances or psychoactive drugs.

  • 2 divider slots allow the zebrafish to choose a chamber or retain the zebrafish in the center area
  • Floor is clear to allow for location preferences
  • Customized floor patterns can be included in your order upon request
  • Sizing modifications can be requested within 50% of the original sizing

In the CPP task, the positive-reinforcing effects of drugs are assessed by the preference of the zebrafish to linger in the environment that was previously conditioned with a substance (drug). The CPP paradigm exploits the natural tendency of the zebrafish to choose black over white. In this assay, the primary stimulus (drug) serves as an unconditioned stimulus. When it is paired with a secondary stimulus (visual cue), which acts as a conditioned stimulus, an approaching behavior for the paired environment is evoked. (Darland & Dowling, 2001).