Orders LinkContactUsLinkComponent
Radial Arm Mazes
Radial Arm Mazes
Supplier:   MAZE ENGINEERS
Radial Arm Mazes
Supplier:  MAZE ENGINEERS
Certificates
Radial Arm Mazes
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The radial arm maze (RAM) is a behavioral test used to assess spatial learning and memory in rodents.
  • Acrylic construction
  • Easy assembly and disssembly with a removable base
  • Easy to clean and easy storage
  • Flexible design of the arms allows for different configurations
The Radial Arm Maze (RAM) consists of a central platform with several arms radiating out from it. It is made of acrylic material. The arms can be baited with food or other rewards. The 3D Radial Arm Maze is a modified version of the Radial Arm Maze developed by Abdel Ennaceur in 2006. Ennaceur’s 3D radial arm maze became a groundbreaking venture because of the unique design; the subjects are exposed to unfamiliar open spaces without a safe alternative.
Product Family Options
Product Information
  • Color
    Length
    Width
    Height
    Availability
    Price
  • 76669-988
    Radial arm maze; mouse
    Radial arm maze; mouse
    Clear, white, grey, black, blue
    35 cm (arm)
    5 cm (arm)
    10 cm (wall)
    Log in for availability
    Log in for availability
    Each
    Each - $3,737.61
    $3,737.61
    Each
       $3,737.61
    Supplier Number: 3901
  • 76669-990
    Radial arm maze; rat
    Radial arm maze; rat
    Clear, white, grey, black, blue
    50 cm (arm)
    10 cm (arm)
    20 cm (wall)
    Log in for availability
    Log in for availability
    Each
    Each - $4,590.94
    $4,590.94
    Each
       $4,590.94
    Supplier Number: 3902
  • 76669-994
    3D radial arm maze; mouse
    3D radial arm maze; mouse
    Log in for availability
    Log in for availability
    Each
    Each - $6,638.95
    $6,638.95
    Each
       $6,638.95
    Supplier Number: 3910
  • 76669-996
    3D radial arm maze; rat
    3D radial arm maze; rat
    Log in for availability
    Log in for availability
    Each
    Each - $7,492.29
    $7,492.29
    Each
       $7,492.29
    Supplier Number: 3911
Accessories
Product Information
  • Catalog #
    Ø
    Availability
    Price
  • Food wells
    Food wells
    76669-992
    2.5 cm
    Log in for availability
    Each - $273.07