MaxiChem® Cut™ Latex Blend Coated Gloves with TriTech™ Liner and Non-Slip Grip on Palm and Fingers
: PIP
MaxiChem® Cut™ cut-resistant latex chemical gloves.
- Chemical resistance delivered through ATG® LiquiTech® technology platform
- Triple-layered TriTech™ technology for added protection, performance, and comfort
- Tailored cuff keeps fingers at the tip of the glove to maximize dexterity
- Micro-cup non-slip grip finish allows for a controlled and superior grip in oily and wet applications
- Non-slip grip is only applied where needed (palm area) to ensure flexibility isn't compromised
Chemical cut gloves feature innovative fibers and base ingredients developed into unique, high-performance yarns and fibers that offer cut protection while maintaining high levels of comfort. An ultra-lightweight synthetic coating, combined with an industry leading super-light seamless knit liner, provides excellent comfort, sensitivity, fit, and dexterity.
: EN 374 results: KLMNOP. ANSI Cut Level: A2. EN 388 2016 results: 4341CX.
- Catalog No:
- 77320-752
- 77320-750
- 77320-754
- 77320-758
- 77320-756
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex
- Latex
- Latex
- Latex
- Latex
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Material:
- Latex
- Latex
- Latex
- Latex
- Latex
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile
Frequently Bought Together