Assurance® Nitrile Coated Gloves with PolyKor® Blended Liner and MicroFinish™ Grip on Palm and Fingers
Supplier: PIP
Certificates
About this item
Assurance® cut-resistant nitrile chemical gloves.
- Nitrile polymer provides excellent grip in wet, oily, and dry conditions
- PolyKor® blended liner is lightweight and provides excellent dexterity, tactile sensitivity, and cut resistance
- MicroFinish™ coating provides excellent resistance to liquid permeation
- Strong protection against punctures and abrasions
Chemical cut gloves feature a MicroFinish™ coating that is infused with thousands of tiny suction cup pockets that create a vacuum effect, dispensing fluids away on contact.
Certifications: Tested to EN 374-5:2016 for protection against micro-organisms. ANSI Cut Level: A4. EN 388 2016 results: 4X42DX. ANSI Abrasion Level: 6. ANSI Puncture Level: 3.
Specifications
- Catalog No:
- 77320-744
- 77320-742
- 77320-748
- 77320-740
- 77320-738
- Ambidextrous/Hand-specific:
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Hand-Specific
- Disposable/Reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Latex-Free
- Powdered/Powder-free:
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Powder-Free
- Material:
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Nitrile
- Sterility:
- Non sterile
- Non sterile
- Non sterile
- Non sterile
- Non sterile