Order Entry
United States
ContactUsLinkComponent
Internal Standard, 4,000 µg/mL in Methylene Chloride for US EPA Methods 625, 8270 and CLP Series Methods, SPEX CertiPrep
Internal Standard, 4,000 µg/mL in Methylene Chloride for US EPA Methods 625, 8270 and CLP Series Methods, SPEX CertiPrep
Catalog # SXCLPS-I
Supplier:  SPEX CERTIPREP LLC
CAS Number:  
undefined
Internal Standard, 4,000 µg/mL in Methylene Chloride for US EPA Methods 625, 8270 and CLP Series Methods, SPEX CertiPrep
Catalog # SXCLPS-I
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  CLPS-I
CAS Number:  

Specifications

  • Concentration:
    4000 µg/ml
  • Matrix:
    Methylene chloride
  • Pack type:
    Ampoule, amber
  • Description:
    Internal Standard
  • Size:
    1 ml
  • Shelf Life:
    36 months
  • Storage Temperature:
    Ambient
  • Cat. No.:
    SXCLPS-I
  • No. of analytes:
    6

Specifications

About this item

Internal Standard for US EPA Methods 625, 8270 and CLP Series Methods, 4,000 µg/mL (4,000 ppm) in Methylene Chloride, 1 mL

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA Methods 625, 8270 and CLP Series Methods
  • 4,000 ppm Internal Standard in methylene chloride matrix

Analytes: Acenaphthene-d10, Chrysene-d12, 1,4-Dichlorobenzene-d4, Naphthalene-d8, Perylene-d12, Phenanthrene-d10

US EPA Method 625 is an analytical method for the monitoring of acid and base neutral extractable fractions in municipal and industrial wastewater by GC/MS.

US EPA Method 8270 is a general analytical method for the monitoring of semivolatiles in environmental and waste samples by GC/MS.

CLP Series Methods: The US EPA retains analytical services through the Contract Laboratory Program (CLP). The CLP follows detailed SOPs derived from EPA methods and SW-846. The CLP Volatile Organic Analysis Method is an analytical method for the analysis of aqueous or solid samples for VOCs by GC/MS.