Eisco Glass Prism and Lens Set, 7-Pieces
470345-096
: Wards
- Description:Glass prism and lens set, 7-pieces
- Cat. No.:470345-096
A variety of lenses and prism block shapes, packed in a wood storage case.
- Ideal for a variety of experiments in optics, light refraction, spectrometry and wavelengths
- Made of highly-polished glass
- Comes in a sturdy, polished wooden storage box with custom foam inserts
Set of 7 optics elements in a wooden storage box. Each of the elements is made of durable, highly polished glass and measures 0.5" (13 mm) thick. Excellent for physics classrooms, as well as photography lighting and distortion effects.
: Kit includes (1) equilateral trianglular prism, 3"; (1) triangular prism, 2×1×0.5"; (1) triangular prism, 2×1×0.75"; (1 ) trapezoid, 1.3" and 1.6" legs; (1) semi-circle, Ø 3.5"; (1) double convex prism, 3.5" with a 3.75" inner radius; (1) double convex prism, 3.5" with a 3.75" outer radius.