Anti-beta Amyloid Mouse Monoclonal Antibody [clone: AB9]
Mouse monoclonal (AB9) antibody to beta Amyloid for WB, ICC/IF and IHC with samples derived from Human.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:1000 to 1:2000, IF: 1:1000, IHC: 1:100
Type: Primary
Antigen: beta Amyloid
Clonality: Monoclonal
Clone: AB9
Conjugation: Unconjugated
Epitope: 1 - 16 aa.
Host: Mouse
Isotype: IgG2a
Reactivity: Human
- Catalog No:
- 77078-750
- 77066-004
- Antigen Symbol:
- beta Amyloid
- beta Amyloid
- Antigen Name:
- A4_HUMAN
- A4_HUMAN
- Antigen Synonyms:
- AICD-59|ABETA|AD1|A4_HUMAN|ABPP|AID(50)|AICD-50|AICD-57|AID(57)|AAA
- AICD-59|ABETA|AD1|A4_HUMAN|ABPP|AID(50)|AICD-50|AICD-57|AID(57)|AAA
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- AB9
- AB9
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P05067
- UniprotID# P05067
- Isotype:
- IgG2a
- IgG2a
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Epitope:
- 1 - 16 aa.
- 1 - 16 aa.
- Immunogen:
- 1-42 human amyloid Aß peptide
- 1-42 human amyloid Aß peptide
- Form:
- Liquid
- Liquid
- Molecular Weight:
- ~5 kDa
- ~5 kDa
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF, IHC
- ICC/IF, IHC
Frequently Bought Together