Anti-Visinin Like Protein 1 Mouse Monoclonal Antibody [clone: 2D11]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal (2D11) antibody to Visinin Like Protein 1 for WB, ICC/IF and IHC with samples derived from Human, Rat and Mouse.
- Validated Applications: WB, ICC/IF, IHC
- Recommended Dilutions: WB: 1:500 to 1:1000, ICC/IF: 1:500 to 1:1000, IHC: 1:500 to 1:100
Type: Primary
Antigen: Visinin Like Protein 1
Clonality: Monoclonal
Clone: 2D11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Rat;Mouse
Specifications
- Catalog No:
- 77066-076
- 77071-938
- Antigen Symbol:
- Visinin Like Protein 1
- Visinin Like Protein 1
- Antigen Name:
- 21 kDa CABP
- 21 kDa CABP
- Antigen Synonyms:
- HPCAL 3|HPCAL3|Hippocalcin-like protein 3|Neural visinin-like type 1 protein|HLP3|Neural visinin-like protein 1|Hippocalcin like protein 3|HLP 3|HUVISL1
- HPCAL 3|HPCAL3|Hippocalcin-like protein 3|Neural visinin-like type 1 protein|HLP3|Neural visinin-like protein 1|Hippocalcin like protein 3|HLP 3|HUVISL1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- 2D11
- 2D11
- Reactivity:
- Human, Rat, Mouse
- Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P62760
- UniprotID# P62760
- Isotype:
- IgG1
- IgG1
- Western Blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Recombinant full-length human Visinin-like Protein 1, expressed in and purified from E. coli
- Recombinant full-length human Visinin-like Protein 1, expressed in and purified from E. coli
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 18 kDa
- 18 kDa
- Purification:
- Immunogen affinity purification.
- Immunogen affinity purification.
- Concentration:
- 1 mg/ml
- 1 mg/ml
- Storage Buffer:
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Supplied in Phosphate buffered saline with 50% Glycerol and 5mM Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Shipped on blue ice at +4 °C
- Tested Applications:
- ICC/IF, IHC
- ICC/IF, IHC
Specifications
Related Information
Frequently Bought Together