Anti-RHCG Goat Polyclonal Antibody
Catalog # 76956-040
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Ammonium transporter Rh type C
- Antigen Symbol:RHCG
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RHCG|PDRC2|RHCG_HUMAN|CDRC2|Rh glycoprotein kidney|chromosome 15 open reading frame 6|C15orf6|Rh family type C glycoprotein|Rh type C glycoprotein
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-ERQNSVYQSD
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human RHCG (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:76956-040
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to RHCG for ELISA, WB and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: RHCG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human