Anti-Cytochrome P450 2E1 Goat Polyclonal Antibody
Catalog # 76955-446
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:4-nitrophenol 2-hydroxylase
- Antigen Symbol:Cytochrome P450 2E1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Cytochrome P450 isozyme 3A|CYPIIE1|Cytochrome P450 2E1|CYP2E|CPE1|CP2E1_HUMAN|Cytochrome P450, family 2, subfamily e, polypeptide 1|Cyp2e1|Cytochrome P450 LM3A
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-RKVIKNVAEVK
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human CYP2E1 (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:76955-446
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to Cytochrome P450 2E1 for ELISA, WB and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: Cytochrome P450 2E1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human