Anti-POU2F1 Goat Polyclonal Antibody
76956-384
- Antibody Type:Primary
- Antigen Name:FLJ42836
- Antigen Symbol:POU2F1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Octamer binding transcription factor 1|NF A1|Octamer-binding protein 1|45200|Oct 1B|NF-A1|Octamer binding protein 1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-NVQSKSNEESGDSQQ
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human POU2F1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:76956-384
- Packaging:Plastic vial
Goat polyclonal antibody to POU2F1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, W
Type: Primary
Antigen: POU2F1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human