Anti-DTX3 Goat Polyclonal Antibody
Catalog # 77075-142
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Deltex3
- Antigen Symbol:DTX3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:RNF154|DTX3|Protein deltex 3|DTX3_HUMAN|Protein deltex-3|RING finger protein 154
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-KNKVTVSKPVWD
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human DTX3 (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77075-142
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to DTX3 for ELISA, WB and IHC with samples derived from Mouse and Human.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: DTX3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse;Human