Anti-PROKR2 Goat Polyclonal Antibody
Catalog # 77066-226
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:B830005M06Rik
- Antigen Symbol:PROKR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:G protein coupled receptor 73 like 1|dJ680N4.3|GPRg2|HH3|G protein coupled receptor I5E|G-protein coupled receptor 73-like 1|GPR73B|EG-VEGRF2|GPR73L1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-NGNTSFTPNFN
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human PROKR2 (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77066-226
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to PROKR2 for ELISA, WB and IHC with samples derived from Human and Rat.
- Validated Applications: ELISA, WB, IH
Type: Primary
Antigen: PROKR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Rat