Anti-FZD2 Goat Polyclonal Antibody
77063-618
- Antibody Type:Primary
- Antigen Name:Frizzled 2
- Antigen Symbol:FZD2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Fz-2|FZD2|FZD 2|Fz2|FZD2_HUMAN|Frizzled-2|Frizzled2|Fz 2|Frizzled homolog 2
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:RYATLEHPFHCPR
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human FZD2 (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77063-618
- Packaging:Plastic vial
Goat polyclonal antibody to FZD2 for ELISA, WB, IF and IHC with samples derived from Human.
- Validated Applications: ELISA, WB, IF, IH
Type: Primary
Antigen: FZD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human