Anti-B MyB Goat Polyclonal Antibody
77063-920
- Antibody Type:Primary
- Antigen Name:B MyB
- Antigen Symbol:B MyB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Rat
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:BMyB|Myb-related protein B|Myb related protein B|MGC15600|MYBB_HUMAN|MybB|B-Myb|MYB L2|Myb-like protein 2
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-QEKARQLLGRLKPSH
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human MYBL2 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77063-920
- Packaging:Plastic vial
Goat polyclonal antibody to B MYB for ELISA and WB with samples derived from Rat.
- Validated Applications: ELISA, W
Type: Primary
Antigen: B MyB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rat