Anti-NPY5R Goat Polyclonal Antibody
Catalog # 77062-098
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Neuropeptide Y receptor type 5
- Antigen Symbol:NPY5R
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Hamster,Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:NPY Y5 receptor|NPY-Y5 receptor|NPY5 R|NPY 5R|NPY Y5|NPY5-R|Neuropeptide Y receptor Y5|NPY5R|NPY R5
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-GLSNKENRLEEN
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human NPY5R (internal region)
- Tested Applications:IHC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77062-098
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to NPY5R for ELISA, WB, IP and IHC with samples derived from Human and Hamster.
- Validated Applications: ELISA, WB, IP, IH
Type: Primary
Antigen: NPY5R
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Hamster