Anti-SH3BP1 Goat Polyclonal Antibody
Catalog # 77067-832
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:SH3 domain-binding protein 1
- Antigen Symbol:SH3BP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:FLJ21318|3BP-1|dJ37E16|SH3 domain binding protein 1|3BP 1|OTTHUMP00000028981|SH3BP1|ARHGAP43|3BP1_HUMAN
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-PQPRPRSLASETN
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human SH3BP1 (C terminal)
- Tested Applications:IHC, ICC
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77067-832
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to SH3BP1 for ELISA, WB, IHC, ICC and IF with samples derived from Human.
- Validated Applications: ELISA, WB, IHC, ICC, I
Type: Primary
Antigen: SH3BP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human