Anti-FSTL1 Goat Polyclonal Antibody
Catalog # 77062-558
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Anti FSTL1
- Antigen Symbol:FSTL1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Follistatin-like protein 1|FRP|FSTL1_HUMAN|FLJ52277|Follistatin like 1|FLJ50214|FSL1|Follistatin related protein|Follistatin-related protein 1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-TAEKTKRVSTKEI
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human FSTL1 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77062-558
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to FSTL1 for ELISA, WB, IF and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, WB, IF, Flow Cytometr
Type: Primary
Antigen: FSTL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human