Anti-IPO7 Goat Polyclonal Antibody
Catalog # 77064-372
Supplier: Avantor
Specifications
- Antibody Type:Primary
- Antigen Name:FLJ14581
- Antigen Symbol:IPO7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:IPO7_HUMAN|Importin-7|MGC138673|IPO7|Imp 7|Importin 7|Importin7|Imp7|IPO 7
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-SSFNFGGPAPGMN
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human IPO7 (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77064-372
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to IPO7 for ELISA and Flow Cytometry with samples derived from Human.
- Validated Applications: ELISA, Flow Cytometr
Type: Primary
Antigen: IPO7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human