Anti-PPP2R5E Goat Polyclonal Antibody
Catalog # 77066-264
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:2A5E_HUMAN
- Antigen Symbol:PPP2R5E
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:PP2A B subunit isoform PR61-epsilon|PP2A B subunit B56 epsilon isoform|PP2A B subunit B' epsilon isoform|PP2A B subunit isoform B56-epsilon|PP2A B subunit isoform B''-epsilon|Epsilon isoform of regulatory subunit B56 protein phosphatase 2A|PP2A B subunit B' epsilon|PP2A B subunit B56 epsilon|PP2A B subunit isoform B'-epsilon
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-LKRGLRRDGIIPT
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human PPP2R5E (C terminal)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77066-264
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to PPP2R5E for ELISA and WB with samples derived from Human, Mouse and Rat.
- Validated Applications: ELISA, W
Type: Primary
Antigen: PPP2R5E
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human;Mouse;Rat