Anti-CLCA1 Goat Polyclonal Antibody
Catalog # 77065-130
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:CaCC
- Antigen Symbol:CLCA1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Calcium dependent chloride channel 1|CaCC-1|CACC1|CLCA family member 1 chloride channel regulator|Chloride channel calcium activated family member 1|Calcium-activated chloride channel family member 1|Calcium-activated chloride channel regulator 1|Calcium-activated chloride channel protein 1|CLCA1
- Storage Buffer:Supplied in Tris Buffered Saline, pH 7.30, with 0.02% Sodium Azide and 0.5% BSA
- Sequence:C-TVTSKTNKDTSK
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Concentration:100 µg at 0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at +4 °C
- Immunogen:Synthetic peptide corresponding to Human CLCA1 (internal region)
- Purification:Purified from goat serum by ammonium sulphate precipitation followed by antigen affinity chromatography using the immunizing peptide.
- Cat. No.:77065-130
- Packaging:Plastic vial
Specifications
About this item
Goat polyclonal antibody to CLCA1 for ELISA and WB with samples derived from Human.
- Validated Applications: ELISA, W
Type: Primary
Antigen: CLCA1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human