Anti-Lamin B1 Rabbit Monoclonal Antibody [clone: ARC0621]
Supplier: Avantor
Certificates
About this item
Rabbit monoclonal [ARC0621] antibody to Lamin B1 for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:20
Type: Primary
Antigen: Lamin B1
Clonality: Monoclonal
Clone: ARC0621
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Frequently Bought Together