Anti-Rag D Rabbit Polyclonal Antibody
Rabbit polyclonal antibody to Rag D for WB and IHC with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 1:1000 to 1:4000, IHC: 1:50 to 1:20
Type: Primary
Antigen: Rag D
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- 76852-420
- Antigen Symbol:
- Rag D
- Antigen Name:
- bA11D8.2.1
- Antigen Synonyms:
- OTTHUMP00000016853|Ras-related GTP binding protein D|Rag D protein|Rag D|DKFZp761H171|FLJ44176|RagDRRAGD|RagD|Ras related GTP binding D
- Antibody Type:
- Primary
- Clonality:
- Polyclonal
- Conjugation:
- Unconjugated
- Reactivity:
- Human, Rat, Mouse
- Host:
- Rabbit
- Gene ID:
- UniprotID# Q9NQL2
- Isotype:
- IgG
- Western Blot:
- Yes
- ImmunoChemistry:
- Yes
- Immunogen:
- Recombinant fusion protein containing a sequence corresponding to amino acids 181-400 of human RRAGD (NP_067067.1)
- Sequence:
- VDGLSDDHKIETQRDIHQRANDDLADAGLEKIHLSFYLTSIYDHSIFEAFSKVVQKLIPQLPTLENLLNIFISNSGIEKAFLFDVVSKIYIATDSTPVDMQTYELCCDMIDVVIDISCIYGLKEDGAGTPYDKESTAIIKLNNTTVLYLKEVTKFLALVCFVREESFERKGLIDYNFHCFRKAIHEVFEVRMKVVKSRKVQNRLQKKKRATPNGTPRVLL
- Form:
- Liquid
- Molecular Weight:
- 46 kDa
- Purification:
- Affinity purification.
- Storage Buffer:
- Supplied in Phosphate buffered saline, pH 7.3, with 50% Glycerol and 0.02% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at +4 °C
- Tested Applications:
- IHC
Frequently Bought Together